Drukuj
Homogenizator, T 10 basic ULTRA-TURRAX®
Dostawca: IKA
3737000 3737002 3737003
431-0179EA 8780 PLN
431-0179 431-0188 431-0189
Homogenizator, T 10 basic ULTRA-TURRAX®
Homogenizatory Homogenizatory ręczne
Handheld or stand model, volume range 0,5 to 100 ml. A wide speed range allows you to work at high circumferential speeds even with small rotor diameters.
  • Quick-release coupling makes changing the dispersing elements easy
  • Excellent speed stability with various media
  • Disposable plastic dispersing elements are available in two sizes
  • These are ideal for PCR analysis
IP protection class according to DIN EN 60529: IP 30
Certyfikaty: IP protection class according to DIN EN 60529: IP 30
Dodatkowe informacje: The homogeniser is supplied with an emptyWięcej informacji o produkcie