Drukuj
dNTP mix
Dostawca: Biotium
40053 40054
40053.EA 1280 PLN
40053. 40054.
dNTP mix
Odczynniki do kwasów nukleinowych Nukleotydy
Highly purified mix of dNTPs (>99% by HPLC) in 0,6 mM Tris-HCl (pH 7,5); each dNTP is at a concentration of 10 or 25 mm in the solution provided.
The dNTPs are suitable for use in PCR, cDNA synthesis, sequencing, and other molecular biology applications.