Drukuj
Szalki Petriego, Sterilin®
Dostawca: STERILIN
504
391-2031EA 4110 PLN
391-2031
Szalki Petriego, Sterilin®
Szalki do hodowli Szalki Petriego
Ideal for use in routine hygiene monitoring of surfaces.
  • Concave profile of the dish raises the profile of the media when set giving better contact between the agar bed and surface under test
  • Aseptically manufactured under cleanroom conditions (class 7 ISO 14644) to exclude microbiological contamination
  • Triple vented to aid gaseous exchange
  • Numbered grid on the base facilitates colony counting
  • Deep skirted base aids stability when stacked
Dimensions shown are for outer Ø base and outer height (mm).
Certyfikaty: Conforms to IES 1993 and IPF 1990 standards.