Drukuj
Heratherm™ Cooled Incubators
Dostawca: Thermo Fisher Scientific
EIS750 EIG750
390-1710EA 90800 PLN
390-1710 390-1711
Heratherm™ Cooled Incubators
Inkubatory Inkubatory z chłodzeniem
Heratherm™ cooled incubators offer exceptional environmental control in a hassle-free chamber that is easy to program and use.
  • Reduce your environmental footprint while achieving outstanding results with Heratherm cooled incubators decreased water and energy consumption and Peltier technology
  • Water-efficient humidification system with integrated water tank and optional clean water connection
  • Peltier technology supports temperature stability and uniformity with lower energy usage compared to traditional compressors
  • Different models available offering
Więcej informacji o produkcie