Drukuj
Próbnik do substancji o średniej lepkości, ViscoSampler
Dostawca: BURKLE
5331-2100 5331-2200 5331-1100 5331-1200 5331-1060 5331-2060
331-2160EA 2640 PLN
331-2160 331-2161 331-2162 331-2163 331-0038 331-0039
Próbnik do substancji o średniej lepkości, ViscoSampler
Próbniki Próbniki do cieczy
ViscoSampler with powerful suction for viscous media. The media is drawn up and discharged using the suction flask.
  • Made of ultra-pure, chemically inert PTFE/FEP for contamination-free sampling
  • Easy cleaning - all surfaces are poreless and without crevices
  • Dirt accumulation is prevented
  • Only round screw threads tried and tested in the field of food hygiene are used
  • Disassembly and cleaning are extremely simple
Specially developed for viscous substances such as slurries, silt, wet clay and soil samples, soil sediments, oils, emulsions and creams.