Drukuj
Chochle z długim uchwytem, wykrywalne, SteriPlast®
Dostawca: BURKLE
5378-5024
331-0361EA 184 PLN
331-0361
Chochle z długim uchwytem, wykrywalne, SteriPlast®
Szufelki do próbek
The ladle is suitable for taking samples of liquids and viscous media as well as powders and granulates. The ladle has a very smooth bottom and rounded corners. This makes it possible to reach into any corner of a container or to take medium directly from the bottom of the container. A long handle with indentations for fingers ensures a firm hold and precise work.
  • PS detectable
  • Individually packaged and sterilised, for single use
  • Reaches into every corner, handle length 300 mm
  • Can be detected by colour scanners, metal detectors or X-rays
  • Can be used as
Więcej informacji o produkcie