Drukuj
Worki z próbnikiem do powierzchni z gąbką celulozową, Whirl-Pak®
Dostawca: NASCO FORT ATKINSON
B01245WA B01422WA B01423WA
300-0185EA 1780 PLN
300-0185 300-0669 300-0670
Worki z próbnikiem do powierzchni z gąbką celulozową, Whirl-Pak®
Worki Woreczki na próbki
This Whirl-Pak® Speci-Sponge® environmental surface sampling bag is designed for environmental surface sampling of work areas, equipment, animal carcasses, and any other place where testing for Listeria, Salmonella, E.coli, coliforms, and other foodborne pathogens or anthrax is required.
  • Available with dry or hydrated sponge
  • Bag can be frozen to any temperature, even with liquid nitrogen, but should not be used at temperatures above 180 °F (82 °C)
  • Wearing a sterile glove when wiping with the sponge is recommended to help prevent contamination
  • Disposable
Więcej informacji o produkcie