Drukuj
PVF torby gazowe i ciekłe do pobierania próbek
Dostawca: Saint Gobain Life Sciences
D1075011
300-0280EA 591 PLN
300-0280
PVF torby gazowe i ciekłe do pobierania próbek
Worki Worki na próbki gazu
Made from 2 mil thick PVF fluoropolymer film. When the chemical inertness of FEP is not required, Chemware® heat-sealed, PVF gas sampling bags are the ideal choice. These reusable bags are the perfect economical alternative for analysis of non-corrosive gases. Can be used when the user is dealing with carbon monoxide, sulphur dioxide, hydrogen sulfide, radon and mercaptans. Ideal for extracting air samples from smoke stack exhaust systems in fuel gas sampling applications.
  • Tough, easy to use, reliable
  • Highly flexible – easy to handle and store
  • Leak proof,
Więcej informacji o produkcie