Drukuj

Anti-IgM Goat Polyclonal Antibody (Biotin)

Dostawca: Tonbo Biosciences
30-8064-M002
30-8064-M002EA 1260 PLN
30-8064-M002
Anti-IgM Goat Polyclonal Antibody (Biotin)
Przeciwciała
Goat anti-Mouse IgM (H&L) - Affinity Pure, Biotin Conjugate

Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: Heavy and light chain
Host: Goat
Isotype:
Reactivity: Mouse
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR