Drukuj
Smar laboratoryjny wolny od silikonu, glisseal N
Dostawca: Borer Chemie
514205.00-D10G
291-1111EA 1870 PLN
291-1111
Smar laboratoryjny wolny od silikonu, glisseal N
Środki nawilżające
Silicone-free laboratory grease for sealing and lubricating fixed and rotary ground glass joints and for greasing fittings made of plastics and metall. Withstands vacuum conditions.
  • Apply only a very thin layer
  • Withstands vacuum conditions
  • Excellent thermal stability, constant lubricating consistency between −40 and +200 °C, in the short run even up to 300 °C
  • No dripping point
  • Chemically stable against most acids, alkalis and gases
  • Silicone-free
  • Can be easily removed with the deconex® cleaning concentrates
  • Technical support from the manufacturer and distributor