Drukuj
Kolumny do chromatografii powinowactwa, HiTrap™ TALON® crude
Dostawca: Cytiva
28-9537-66 28-9537-67 29-0485-65
28-9537-66EA 1220 PLN
28-9537-66 28-9537-67 29-0485-65
Kolumny do chromatografii powinowactwa, HiTrap™ TALON® crude
Kolumny chromatograficzne
HiTrap TALON® crude 1 and 5 ml columns allow for rapid separations and easy scale-up with a minimum of sample preparation and equipment.
  • Columns prepacked with cobalt-based chromatography media designed for purification of histidine-tagged recombinant proteins by immobilized metal affinity chromatography (IMAC)
  • Suitable for IMAC purification of histidine-tagged proteins when Ni²⁺ is not the optimal choice of metal ion
  • Compatible with commonly used IMAC reagents and appropriate for purifying proteins under native or denaturing conditions
  • Prepacked HiTrap
Więcej informacji o produkcie