Drukuj
Folia, Amersham Hyperfilm™ MP
Dostawca: Cytiva
28-9068-50 28-9068-46 28-9068-45 28-9068-44 28-9068-43 28-9068-42
28-9068-50EA 1810 PLN
28-9068-50 28-9068-46 28-9068-45 28-9068-44 28-9068-43 28-9068-42
Folia, Amersham Hyperfilm™ MP
Filmy do autoradiografii Filmy
Amersham Hyperfilm™ MP is a multipurpose film constructed from a clear base coated on both sides with a photographic emulsion that is protected by an anti-scratch layer. The anti-scratch coating reduces the risk of visible artefacts (e.g., plastic wrap marks) and the clear colourless base results in easy reading of images, without compromising on sensitivity.
  • High sensitivity for low concentration targets
  • Intended for radiometric or chemiluminescent applications
  • Clear background for excellent contrast and improved band visibility
  • Publication quality images
Więcej informacji o produkcie