Drukuj
Smary próżniowe, Apiezon®
Dostawca: M I MATERIALS
FAP/G/L-00025 FAP/G/L-00050 FAP/G/M-00025 FAP/G/M-00100 FAP/G/H-00025 FAPGAP10104000 AP100-0050
24486.135EA 1670 PLN
24486.135 24486.157 24487.138 24487.184 24491.133 MIMAFAPGAP10104000 MIMAAP100-0050
Smary próżniowe, Apiezon®
Środki nawilżające
These Apiezon greases are hydrocarbon based, silicone-free and highly resistant to creep, the undesirable phenomenon associated with silicone-based products.
  • Excellent performance in a wide range of vacuum applications
  • Silicone-free, highly creep resistant
  • Solutions for a variety of temperatures
  • Stable, chemically inert and non toxic
  • Easy to use and remove
Using Apiezon reduces the chance of grease travelling away from the area of application and contaminating adjacent surfaces. In addition the ultra high vacuum grease ensures ultimate cleanliness andWięcej informacji o produkcie