Capillary columns, traditional phases, SUPELCOWAX® 10, Supelco®
Supplier: Merck
24080-U 24084 25375-U 24169 24211 24212 24284 24343 25300-U 25301-U 25325 25376 23308-U 25391 24077 24078 24079 24081 24082 24085-U
24080-UEA 4820 PLN
24080-U 24084. 25375-U 24169. 24211. 24212. 24284. 24343. 25300-U 25301-U 25325. 25376. 23308-U 25391. 24077. 24078. 24079. 24081. 24082. 24085-U
Capillary columns, traditional phases, SUPELCOWAX® 10, Supelco®
Chromatography Columns
These bonded SUPELCOWAX® 10 columns are widely used for separation and purity analyses of many polar compounds, including alcohols, aromatics, and other solvents, flavors, fragrances and FAMEs.
  • High thermal stability
  • Temperature limits from 35 to 280 °C
Certifications: Meets USP G16 requirements.
Order Now