Drukuj
Nożyczki, Sauro, Rubis®
Dostawca: OUTILS RUBIS
1F007/EL
233-0225EA 213 PLN
233-0225
Nożyczki, Sauro, Rubis®
Nożyce
Micro scissors with wider and longer steel profile to ensure better strength when cutting hard tissues.
  • Produced from hardened stainless steel AISI420
  • Highly resistant against acids and alkaline
  • With ergonomically designed Rubis Twist® grips
  • No Medical Device (MD) product
Made in Switzerland to the highest quality standards. Rubis products are the result of traditional craftsmanship, strict throughput controls and a finishing process under the magnifying glass. No tool leaves the factory without having passed the strict quality controls. NeitherWięcej informacji o produkcie