Drukuj
Pincety wagowe
Dostawca: BOCHEM
1740
232-0061EA 66.8 PLN
232-0061
Pincety wagowe
Pincety
Stainless steel 18/10, antimagnetic, with plastic tips.
  • Ideal for picking up and handling precision weights
  • Max. temperature 145 °C