Drukuj
Szczypce do zlewek, stal nierdzewna
Dostawca: RSG
043.330
231-4145EA 280 PLN
231-4145
Szczypce do zlewek, stal nierdzewna
Stainless steel ¹⁸/₁₀, anti-magnetic. For easy, safe handling of beakers and similar containers. The jaws are wrapped in ceramic fibres to provide thermal insulation and to protect the containers they hold.
  • Corrosion resistant
  • Excellent thermal resistance, autoclavable