Drukuj
Folie uszczelniające, Seal-R-film™
Dostawca: Brand
701620 701624 701626 701622
219-0000EA 257 PLN
219-0000 219-0002 219-0003 219-0001
Folie uszczelniające, Seal-R-film™
Plomby Filmy i folie uszczelniające
Seal-R-film™ sealing film seals beakers, flasks, and other vessels.
  • Protects against evaporation, leakage, and contamination
  • Easy sealing with 300% elasticity
  • Lot identification of each roll
The sealing film can be stretched to three times its original length. This means that Seal-R-film™ always fits, even with irregular shapes. For traceability, each roll has a lot identification. With the BRAND cutter for sealing film you can cut rolls with widths of 50 and 100 mm efficiently and economically and store them cleanly.
Specifications:
- Thickness: 0Więcej informacji o produkcie