Drukuj
VWR®, Karboje, wąska szyjka, okrągłe, z korkiem i zakrętką
Dostawca: VWR Collection
216-1731EA 343 PLN
216-1731 216-1710 216-1717 216-1705 216-1734 216-1718 216-1899 216-1909 216-1928
VWR®, Karboje, wąska szyjka, okrągłe, z korkiem i zakrętką
Butelki i butle Butle typu carboy
LDPE/HDPE or PP, translucent with purple screw caps. Use for storing and dispensing solutions and media. Round carboys feature an integrated shoulder handle. Graduated to identify the volume dispensed.
LDPE carboys
Ideal for high-purity applications as they have high resistance to chemicals (e.g. widely used in ophthalmic product range, like eye droppers and nasal sprays)
Ideal resin for cold storage (down to −80 °C), working temperature −80 to +80 °C
Resistant to physical impact (will not break even if it falls down accidentally, whereas PP carboys may lead to cracking at negativeWięcej informacji o produkcie