Drukuj
Pojedyncze kasety do wafli, do wafli o rozmiarze 4" (100 mm), seria H 22/H 93
Dostawca: ENTEGRIS
H9340-101-1116 H22-402-0615 H22-402A-0615 H22-401-62C02 H22-402-62C02 H22-401-0615 H22-40-0615_U H22-40-62C02
216-1589EA 0 PLN
216-1589 216-2346 216-2349 216-2345 216-2348 216-2344 ENTEH22-40-0615_U 216-2350
Pojedyncze kasety do wafli, do wafli o rozmiarze 4" (100 mm), seria H 22/H 93
Urządzenia do wafli Kasety pojedyncze do wafli
H 22 Series
Conical pocket holds one wafer facedown contacting only the wafer’s edges.
  • Spring retains wafer and minimises particle generation
  • Stackable for efficient use of storage space
  • Loose or lock stacking, either tray on tray or cover on tray
  • Convenient screw on covers
  • Large area on cover provides space for labeling
H 93 Series
Holds one wafer face down contacting only the edge of the wafer. Contains four inserted cushions to retain wafer and minimise particle retention.
Tweezer slots on shipper bottom ease wafer removal
Stackable for efficient useWięcej informacji o produkcie