Drukuj
ThermoSafe® EPS Insulated Shippers with Outer Corrugate
Dostawca: Sonoco Thermosafe
50002062C 50004243C
216-1328EA 2630 PLN
216-1328 216-1336
ThermoSafe® EPS Insulated Shippers with Outer Corrugate
Koperty i pojemniki wysyłkowe
EPS is an effective insulator which protects cargo from external changes in temperature.
  • High-quality EPS base and lid
  • Strong outer corrugate box for extra protection
  • Ideal for the storage and transportation of temperature sensitive product
  • Use in combination with refrigerants or dry ice to create a bespoke solution for your application
EPS insulated containers offer the ideal solution for the safe storage and delivery of many refrigerated or frozen materials. They are lightweight yet tough, making them a popular choice when shipping temperatureWięcej informacji o produkcie