Drukuj
VWR®, Szkiełka zegarkowe
Dostawca: VWR Collection
216-0163EA 154 PLN
216-0163 216-0155 216-0156 216-0157 216-0158 216-0159 216-0160 216-0161 216-0162 216-0454 216-0455
VWR®, Szkiełka zegarkowe
Szkiełka zegarkowe
PTFE.
  • Excellent chemical resistance
  • Temperature resistant from –200 to 280 °C
  • Exceptionally smooth surfaces to prevent contamination
Ideal for covering PTFE beakers when decomposing and evaporating, can also be used as evaporating dishes. Extremely robust.