Drukuj
VWR® Bottles, Wide Neck, Square, with Cap
Dostawca: VWR Collection
215-4550EA 70.5 PLN
215-4550 215-4551 215-4552 215-4553 215-4554 215-4555 215-4556 215-4557 215-4558 215-4559 215-4560 215-4561 215-4562 215-4563 215-4564
VWR® Bottles, Wide Neck, Square, with Cap
Butelki i butle Butle plastikowe
Wide neck square bottles available in HDPE or PP material.
  • Amber color HDPE bottle protects light sensitive materials
  • PP closure ensures leak-proof sealing
  • Square shape is ideal for storage of dry chemicals or powder
Resin compliance to US FDA and US and EU Pharmacopiea.
PP bottles are autoclavable.