Drukuj
Systemy przenoszenia butelek, VWR
Dostawca: DWK Life Sciences
292807309 292826306 292836307 292816305 292806304
215-4172EA 609 PLN
215-4172 215-4170 215-4171 215-4169 215-4168
Systemy przenoszenia butelek, VWR
Pojemniki transportowe Nosidła do butli
This modular bottle carrying system fits standard laboratory bottles that have a GL 45 neck finish.
  • Compatible with four bottle sizes 2, 5, 10 and 20 litres
  • All materials are fully autoclavable
  • Makes pouring liquids from the larger bottles more comfortable and safer
It is ergonomically designed and offers health and safety benefits to the user. It allows easy and safe transport of bottles around the laboratory or production facility.