Drukuj
Kriopojemniki, PP, seria D40/D60/D70/D90, GLW
Dostawca: GLW
D40 D40B D40G D40R D40Y D40S DS40 D70 D70B D70G D70R D70Y D70S DS70 D90 D90B D90G D90R D90Y D90S DS90 D60 D60B D60G D60R D60S D60Y DS60
211-0525EA 19.7 PLN
211-0525 211-0526 211-0527 211-0528 211-0530 211-0529 479-1513 GLWAD70 GLWAD70B GLWAD70G GLWAD70R GLWAD70Y GLWAD70S GLWADS70 211-0531 211-0532 211-0533 211-0534 211-0536 211-0535 479-1566 GLWAD60 GLWAD60B GLWAD60G GLWAD60R GLWAD60S GLWAD60Y GLWADS60
Kriopojemniki, PP, seria D40/D60/D70/D90, GLW
Pudełka Kriopudełka
Cryoboxes, PP, natural or coloured, without divider.
  • Autoclavable up to +121 °C and use in freezer down to –90 °C
  • The included clear lid offers a view of the contents without opening the cryobox
  • Without drainage holes in base for catching drips or store fluids (leakproof only at bottom up to 40 mm)
  • For light sensitive samples please choose the colour Black/black
The boxes comes without compartments, in addition they can be used with GLW dividers for individual configuration.They are suitable to store non standardised tubes, stone samples, small itemsWięcej informacji o produkcie