Drukuj

Anti-IgG Llama Polyclonal Antibody (CF™ 568)

Dostawca: Biotium
20455-50UL 20455-500UL
20455-50ULEA 647 PLN
20455-50UL 20455-500UL
Anti-IgG Llama Polyclonal Antibody (CF™ 568)
Przeciwciała
Llama anti-mouse CF™ dye conjugates are prepared by labeling affinity purified llama anti-mouse IgG (H+L) with a selection of CF™ dyes. CF™ dyes offer combined advantages in brightness, photostability, and specificity compared other fluorescent dyes. Red fluorescent CF™568 (Ex/Em 562/583 nm) produces much brighter antibody conjugates than Alexa Fluor® 568, and has superior photostability.

Alexa Fluor® is a registered trademark of Thermo Fisher Scientific.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF568
Epitope:
Host: Llama
Isotype:
Reactivity: Mouse
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR