Drukuj
Rurki szklane
Dostawca: GLASWARENFABRIK KARL HECHT
92260204055
201-0539EA 111 PLN
201-0539
Rurki szklane
Przewody, kapilary i rury Przewody rurowe
AR-Glass®, clear.
  • Good chemical and physical properties
  • Suitable for the production of items that must normally withstand only brief exposure to aggressive chemicals
  • Not resistant to high thermal loads
Tube length ~750 mm.