Drukuj

Bufor wodorowęglanu trietyloamonu, 1 M, bufor lotny do HPLC, Fluka™

Dostawca: Honeywell Chemicals

17902-100ML 17902-500ML
17902-100MLEA 610 PLN
17902-100ML 17902-500ML
Bufor wodorowęglanu trietyloamonu, 1 M, bufor lotny do HPLC, Fluka™
Bufory Bufory do chromatografii

Triethylammonium bicarbonate buffers are volatile buffer for HPLC.


  • Non-volatile matter: max. 0,05 %
  • pH (20 °C): 8,4 to 8,6
  • Absorbance at 260 nm: Max. 0,04
  • Absorbance at 280 nm: Max. 0,04

Zobacz ceny


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR