Drukuj
Wkłady do dejonizacji wody, Aquadem® DL
Dostawca: ENVIROFALK PHARMAWATERSYSTEMS
1941-005 1941-015 1941-025
172-4109EA 0 PLN
172-4109 172-4111 172-4112
Wkłady do dejonizacji wody, Aquadem® DL
Systemy oczyszczania wody Dejonizatory
Pressureless water deionisation cartridge with plastic housing, filled with regenerable mixed-bed resins.
  • Compact and inexpensive solution for smaller requirements, ready for connection
  • Achieved water quality covers the majority of common laboratory applications
  • Simple handling with quick connection system
  • Suitable for benchtop set up in laboratories
  • Long life time, transportable
SB connection set: Supply and discharge hose, flow-through electrode, outlet tube, SB conductivity meter.
GS connection set: Supply and discharge hose, flow-throughWięcej informacji o produkcie