Drukuj
VWR®, system oczyszczania wody, seria E
Dostawca: VWR Collection
171-1251EA 49300 PLN
171-1251 171-1252 171-1253 171-1473 171-1474 171-1475
VWR®, system oczyszczania wody, seria E
Systemy oczyszczania wody Systemy czystej i ultra czystej wody
This compact system produces EDI-based Type II water directly from tap water. Quality of Type II water meets or exceeds water quality as defined by ASTM, CAP, CLSI and ISO 3696 / BS 3997, and also complies with the purified water requirements from the European and U.S. Pharmacopoeia.
  • Simple and intuitive to dispense water via the dispenser or central console
  • Central console is an 8 inch waterproof touch screen, providing user full control and access of system performance
  • One water system can drive several dispensing units ('1 + N mode')
  • Dispensing flow