This compact system produces EDI-based Type II water directly from tap water. Quality of Type II water meets or exceeds water quality as defined by ASTM, CAP, CLSI and ISO 3696 / BS 3997, and also complies with the purified water requirements from the European and U.S. Pharmacopoeia.
- Simple and intuitive to dispense water via the dispenser or central console
- Central console is an 8 inch waterproof touch screen, providing user full control and access of system performance
- One water system can drive several dispensing units ('1 + N mode')
- Dispensing flow rate is up to 2 L/min and volumetric dispensing is available
- RFID-tagged consumables for easier tracking of lifetime and performance
- Wireless communication option to control multiple dispensers and the central console (optional)
- Real time TOC and resistivity/conductivity monitoring
- Optional tank circulation mode to maintain a stable water quality in the tank
- Users can see essential information (water quality, system status, warnings) on the dispenser unit and the central console at any time (data stored up to 2 years; downloadable)
The graphic display guides users through system operation, performance, troubleshooting and maintenance, and can be password protected for multiple access levels.
Applications: Preparation of chemical and bio-reagents; preparation of culture media; preparation of solutions for chemical analysis (such as HPLC and ICP); clinical analysers; medical devices and equipment rinsing; serum and blood fractionation; ophthalmics.
Certyfikaty: The VWR® water system is delivered with a Certificate of Conformity ensuring that it has been built and tested fully assembled following Standard Operating Procedures, and a Certificate of Calibration for the temperature and resistivity meters built into the system. Consumables are automatically delivered with a Certificate of Quality. Manufacturing site is ISO 9001 (2015) and ISO 140001 certified.
Informacje do zamówienia: For a complete system, the production unit and the mandatory accessories have to be ordered. Other accessories and consumables, including wall mounting brackets, purification cartridges and wireless upgrade kit are available separately.
Please note: For the E5 system you need to order one unit of the RO pack 171-1276 (12 L/h), For the E10 system two units of the RO pack are required 171-1276 (12 L/h). For the E15 system please order two units of the RO pack 171-1277 (16 L/h).
Dodatkowe informacje: VWR® E5 System starter setup (171-1473) includes 1x VWR® E5 System, 1x 0,2 µm PES final filter, 1x VWR® AC Pack, 1x VWR® P Pack, 1x VWR® RO Pack, 12 L/H, 1 x VWR® Cleaning Pack, 1x leak protector, 1x dispenser kit, 1x tank vent filter, 1x UV sanitisation module for VWR® tank, and 1x 30 L PE tank with sensors.
VWR® E10 System starter setup (171-1474) includes 1x VWR® E10 System, 1x 0,2 µm PES final filter, 1x VWR® AC Pack , 1x VWR® P Pack, 2x VWR® RO Pack, 12 L/H, 1x VWR® Cleaning Pack, 1x leak protector, 1x dispenser kit, 1 x tank vent filter, 1x UV sanitisation module, and 1x 30 L PE tank with sensors.
VWR® E15 System starter setup (171-1475) includes 1x VWR® E15 System, 1x 0,2 µm PES final filter for VWR®, 1x VWR® AC Pack , 1x VWR® P Pack , 2x VWR® RO Pack, 16 L/H, 1x VWR® Cleaning Pack, 1x leak protector, 1 x Dispenser kit, 1x tank vent filter, 1x UV sanitisation module , and 1x 30 L PE tank with sensors.
Mniej informacji