Drukuj
Dejonizatory, Micromeg
Dostawca: ELGA LabWater
M/M/DS/A
171-0484EA 0 PLN
171-0484
Dejonizatory, Micromeg
Systemy oczyszczania wody Dejonizatory
This wall mounted disposable deioniser produces water up to a quality of 10M.-cm at flows of up to 90 l/hr. Designed for users with a requirement for low volumes deionised water, the Micromeg requires no electrical input as its water quality meter is battery operated.
  • Disposable cartridge
  • New cartridge can be installed in seconds