Drukuj
Prefabrykowany ExcelGel do SDS-PAGE
Dostawca: Cytiva
17-1342-01
17-1342-01EA 864 PLN
17-1342-01
Prefabrykowany ExcelGel do SDS-PAGE
Elektroforezy, Żele Żele prefabrykaty
ExcelGel precast gels for SDS-PAGE are available as homogeneous or gradient gels. The homogeneous gels have 25 preformed sample wells for sample volumes from 5 to 17 µl. The gradient gels use sample application strips (26 samples up to 40 µl, 52 samples up to 20 µl), paper pieces or sample droplets. All ExcelGel SDS homogeneous and 8 - 18 gradient gels have a 33 mm stacking zone and a 77 mm separating zone.
  • Save time with ready-to-use, mechanically stable gels cast on plastic supports
  • Apply samples quickly and easily using preformed sample wells
  • Avoid