Drukuj
Matryca bazowa do filtracji żelowej, Sepharose™ 4B
Dostawca: Cytiva
17-0120-01
17-0120-01EA 5690 PLN
17-0120-01
Matryca bazowa do filtracji żelowej, Sepharose™ 4B
Systemy oczyszczania białek Protein Purification Resins
Sepharose™ 4B is an agarose gel filtration base matrix and is frequently used for coupling affinity ligands to the matrix. The matrix is not pre-activated and the user performs all steps in coupling.
  • 4% agarose gel filtration media
  • Proven base matrix for coupling affinity ligands