Drukuj

Woda z 0,1% (v/v) Kwas mrówkowy, CHROMASOLV™ LC-MS Ultra LabReady™, Solvent Blend tested for UHPLC-MS, Riedel-de Haën™

Dostawca: Honeywell Chemicals

14281-2L
14281-2LEA 712 PLN
14281-2L
Woda z 0,1% (v/v) Kwas mrówkowy, CHROMASOLV™ LC-MS Ultra LabReady™, Solvent Blend tested for UHPLC-MS, Riedel-de Haën™
Woda z 0,1% (v/v) Kwas mrówkowy

  • For the most demanding LC-MS and UHPLC applications
  • Tested for UHPLC-MS suitability
  • Highest solvent purity for low detection limits
  • Extremely low MS baseline noise

Gęstość: 0.998 g/cm³ (20 °C)
Temperatura przechowywania: Temperatura otoczenia
Numer MDL: MFCD00011332
Numer WE (dawny EINECS): 231-791-2

Zobacz ceny

Wyniki testów specyfikacyjnych

Assay of formic acid 0.093 - 0.107 %
pH (20 °C) 2.6 - 2.8
Calcium (Ca) Max. 0.2 ppm
Potassium (K) Max. 0.2 ppm
Magnesium (Mg) Max. 0.2 ppm
Sodium (Na) Max. 0.5 ppm
Transmittance at 210 nm Min. 5 %
Transmittance at 230 nm Min. 45 %
Transmittance at 254 nm Min. 98.5 %
UHPLC-MS ESI(+) (Reserpine) Max. 10 ppb
UHPLC-MS ESI(-) (Digoxin) Max. 50 ppb
HPLC Gradient at 210 nm Max. 20 mAU
HPLC Gradient at 254 nm Max. 5 mAU
Suitability for the UHPLC-MS complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR