Drukuj

Phosphorus standard dissolved w oil, 1000 µg/g P w mineral oil Specpure® for wear metal analysis

Dostawca: Thermo Fisher Scientific

14210.18
14210.18EA 808 PLN
14210.18
Phosphorus standard dissolved w oil, 1000 µg/g P w mineral oil Specpure® for wear metal analysis
Standardy Wzorce kationowe Phosphorus Standards

Physical Form: Liquid
MDL No.: MFCD00133771

Zobacz ceny


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR