Drukuj
Heavy-Duty 50 GSM SMS Autoclavable Bowl Covers with Steam Indicator
Dostawca: KEYSTONE CLEANROOM PRODUCTS
BWLKCRP30030 BWLKCRP30040 BWLKCRP30036 BWLKCRP30048 BWLKCRP30020 BWLKCRP30042 BWLKCRP30050 BWLKCRP30054 BWLKCRP30024 BWLKCRP30060
129-4013EA 24250 PLN
129-4013 129-4019 KACCBWLKCRP30036 KACCBWLKCRP30048 KACCBWLKCRP30020 KACCBWLKCRP30042 KACCBWLKCRP30050 KACCBWLKCRP30054 KACCBWLKCRP30024 KACCBWLKCRP30060
Heavy-Duty 50 GSM SMS Autoclavable Bowl Covers with Steam Indicator
Pokrywy
This 50 g/m² SMS autoclavable bowl cover provides a microbial barrier that is ideal for pharmaceutical production applications.
  • Hydrophobic, tear-resistant, and puncture-resistant
  • Creates secondary level of protection during aseptic presentation and setup of equipment
  • Designed for autoclave cycles (121 to 135 °C)
  • Manufactured and packed in a certified ISO 8 cleanroom
  • Made in USA
Eliminate the need for cutting sheets of sterilisation wrapping materials and securing them with autoclave tape and rubber bands. After the outer bag is removed, thisWięcej informacji o produkcie