Drukuj
Autoklawowalne osłony na mufy, Tyvek®
Dostawca: KEYSTONE CLEANROOM PRODUCTS
MFT2A03 MFT2A04 MFT2A05 MFT2A05OT MFT2A06BT MFT2A07 MFT2A08 MFT2A09 MFT2A12 MFT2A16 MFT2A18 1034TW
129-3076EA 12440 PLN
129-3076 129-3077 129-3078 129-3123 129-3125 129-3126 129-3128 129-3129- 129-3131 129-3133 129-3134 129-4200
Autoklawowalne osłony na mufy, Tyvek®
Pokrywy
Save time and standardise the activities in equipment preparation for sterilisation.
  • Manufactured and packaged in a controlled cleanroom environment
  • Reduces labour costs
  • Simplifies the conformance to wrapping SOP's
  • Steam sterilisation indicator - autoclavable at 121 to 125 °C
  • Tear- and puncture-resistant, hydrophobic and lint-free Tyvek® 1422A
  • With elastic to fit a range of diameters
  • Conveniently packaged 100 per sealed poly bag
Tyvek® Muffs provide a convenient means to cover flasks, beakers, containers, media bottles, valves, pipes, and hoses. TheseWięcej informacji o produkcie