Drukuj
Sterylne worki na próbki z polem do opisu, Whirl-Pak®
Dostawca: NASCO FORT ATKINSON
B01067WA B01064WA B01062WA B01065WA B01297WA B01195WA B01196WA B01447WA B01446WA B01515WA B01445WA B01341WA B01491WA
NASCB01067WAEA 934 PLN
NASCB01067WA NASCB01064WA NASCB01062WA 129-0669 129-0671 129-0673 129-0674 NASCB01447WA NASCB01446WA NASCB01515WA NASCB01445WA NASCB01341WA NASCB01491WA
Sterylne worki na próbki z polem do opisu, Whirl-Pak®
Worki Woreczki na próbki
Blended, virgin polyethylene film with Write-On strip provides exceptional clarity, superior strength, and sterilised after manufacturing for results you can trust. HACCP, EPA, USDA, FDA approved for food contact.
  • Plus 1 sterilisation, post-manufacturing sterilisation: Increased sample integrity, perforated closure to maintain sterility
  • Convenient write-on strip for easy labeling
  • Puncture-proof closure tabs: Inner and outer tapes for longevity and stability, wire for easy opening, pouring, and closing
  • Opening pull tabs: For easy opening, non-slip textured
Więcej informacji o produkcie
Zobacz ceny
SPECYFIKACJE
Environmentally Preferable