Drukuj
Zestaw do usuwania rozlanych chemikaliów, opakowanie zbiorcze PIG®
Dostawca: NEW PIG
KIT302 KITR302 KIT202-999 KIT402 RFL402
121-0509EA 5170 PLN
121-0509 121-0510 121-0155 121-0563 121-0158
Zestaw do usuwania rozlanych chemikaliów, opakowanie zbiorcze PIG®
Materiały do kontroli wycieków
Spill kits containing all the absorbents needed to respond to a large spill of oils, coolants, solvents or water - all in one container that can then be used to dispose of the waste.
  • Leakproof, twist-on lid is notched so can easily be removed or tightened
  • Overpack is X-rated for Packing Groups I, II and III, so it's certified for shipping waste by land, sea or rail after cleanup is finished
  • Lift-out, prepacked baskets speed access and guard contents from UV
  • Ledges moulded into container make kit easy to move by forklift
  • Lightweight, 100% polyethylene
Więcej informacji o produkcie