Drukuj
Rolki PIG® Rip-&-Fit® HazMat
Dostawca: NEW PIG
MAT342 MAT343
121-0530EA 485 PLN
121-0530 NEWPMAT343
Rolki PIG® Rip-&-Fit® HazMat
Materiały do kontroli wycieków
Extra perforations let you create custom sizes of PIG HazMat Mat without cutting. Just "rip" it along the easy-tear perfs and "fit" it into place.
  • Absorbs the widest range of acids, caustics and unknown liquids - even those with high concentrations like 98% sulfuric acid and 30% sodium hydroxide
  • Chemical-resistant mat won't degrade or cause a dangerous reaction upon contact with corrosive spills
  • Extra perforations in our Rip-&-Fit Mat allow you to easily "rip" this PIG Mat and "fit" it to your application without cutting
  • No matter how you rip it, this
Więcej informacji o produkcie