Drukuj
Pokrowiec na stację roboczą ESD
Dostawca: DESCO
41401
115-3173EA 328 PLN
115-3173
Pokrowiec na stację roboczą ESD
Ochraniacze powierzchni Ochraniacze powierzchni
ESD workstation cover protects the workstation from ESD, dust and other contaminants.
  • Groundable workstation cover
  • Surface resistance: 1×10E5 < 1×10E7 Rtt
  • Minimum of 9% carbon conductive nylon fibers with bound edges
  • 10 mm (³/₈") grounding socket
  • Made in the United States of America
Cover attenuates electrostatic fields and electrostatic discharges (ESD). Minimum of 9% carbon conductive nylon fibres with bound edges provide continuous and consistent charge dissipation throughout lifetime of cover.
Grounding socket (10 mm) allows cover to beWięcej informacji o produkcie