Drukuj
Środek dezynfekujący, sterylny 70% izopropanolu, Contec
Dostawca: CONTEC
SBC570I SBT170IW SBT0570IW
CONTSBC570IEA 1300 PLN
CONTSBC570I 141-1463 115-2473
Środek dezynfekujący, sterylny 70% izopropanolu, Contec
Środki czystości Środki do dezynfekcji powierzchni
Contec® Sterile IPA is a blend of 70% v/v Isopropanol with 30% water for injection or purified water, this ready to use formulation eliminates mixing, filtration, processing and QC expense of in-house production. Used for wiping gloved hands and also as surface cleaner for removal of contaminants particularly when used in conjunction with Contec® sterile wipes.
  • Guaranteed endotoxin levels of <0,25 EU/ml make it ideal for use in product contact areas
  • GMP manufactured under Grade A laminar flow air flow in a Grade C cleanroom ensures the alcohol,