Drukuj
Chusteczki do mycia rąk, wstępnie nasączone, Kresto® Special WIPES
Dostawca: Deb-STOKO
ULT150W ULT70W
115-2381EA 857 PLN
115-2381 115-2384
Chusteczki do mycia rąk, wstępnie nasączone, Kresto® Special WIPES
Chusteczki Chusteczki i ręczniki jednorazowe Chusteczki nasączone
Tough, extra large wipes impregnated with a mild ester solvent-based liquid formula for highly effective removal of paints, inks, resins, adhesives and most other print and paint shop contaminants. For use without water in industrial environments. Suitable for use on-site and for mobile workers.
  • No water needed: Impregnated with an advanced hi-tech formulation solvent liquid that includes dibasic esters
  • Wipes are strong and absorbent, acting quickly and effectively leaving no sticky feeling
  • Highly effective and environmentally friendly moisturiser for
Więcej informacji o produkcie