Drukuj
Detergent do szkła i laboratorium, NovaClean™
Dostawca: MICRONOVA
NC2-G
115-0291EA 675 PLN
115-0291
Detergent do szkła i laboratorium, NovaClean™
Środki czystości Detergenty i środki myjące
Ready to use, low-ionic detergent that can be applied directly to cleanroom work surfaces including glass, stainless steel, epoxy acrylic, and vinyl curtains.
  • Ready to use, cleanroom surfactant
  • Can be applied directly to a range of surfaces
  • Works well with glass, stainless steel, epoxy, acrylic, and vinyl curtains
  • Filtered to 0,1 µm; free of mobile ions
  • Low Ionic; low in sodium and potassium
  • Low non-volatile residue levels
  • Free-rinsing
The Micronova NovaClean™ Lab and Glass Cleaner is a low-ionic detergent, designed for use on cleanroom work-Więcej informacji o produkcie