Drukuj
Paski nasadek, płaska góra
Dostawca: HONEYWELL SAFETY
4503001-XXXL
113-8676EA 0 PLN
113-8676
Paski nasadek, płaska góra
Odzież jednorazowa
Disposable, liquidtight overalls made from three-layered PE for excellent protection against liquids (chemicals, oils, hydrocarbons).
  • Double adhesive flap and welded zipper
  • Excellent abrasion, tear and puncture resistance
  • Watertight due to 'Spaseal' welding
  • Glued gloves and integrated overboots for an increased safety
  • Yellow colour for improved visibility
Thickness: 100 µm
Colour: yellow
Certyfikaty: Type 3/5, EN 340, EN 1511, Cat. III