Drukuj
Kombinezon, PP
Dostawca: HUBEI ORIENT INTL TRADING
113-8104EA 1340 PLN
113-8104
Kombinezon, PP
Odzież jednorazowa
These disposable overalls made from non woven PP provide protection against dust and dirt.
  • With hood
  • Zip closure
  • Elasticated wrist and ankles
  • Single bagged
Suitable for use in low risk areas, such as food processing, manufacturing and hygiene areas. Ideal for visitors.
Weight: 52 g/m²
*One size fits all