Drukuj
Okulary ochronne, uvex i-guard+ planet
Dostawca: UVEX
9143.297
112-6183EA 171 PLN
112-6183
Okulary ochronne, uvex i-guard+ planet
Gogle
These metal-free, versatile 3-in-1 protective eyewear range allows a flexible choice for many applications.
  • Large, flat lens design for an unrestricted field of vision
  • UV400 – 100% protection against dangerous UVA and UVB radiation to 400 nm
  • Sustainably produced using recycled and bio-based materials
  • Individually adjustable headband for a perfect fit and secure hold
The robust polycarbonate lens with proven uvex supravision coating technology with permanent anti-fog and scratch-resistant features. x-tended sideshield provides additional sideWięcej informacji o produkcie