Drukuj
Rękawice nitrylowe odporne chemicznie, PowerCoat® 950-20 Neofit
Dostawca: HONEYWELL SAFETY
2095020/7 2095020/8 2095020/9 2095020/10 2095020/11
112-4040EA 39 PLN
112-4040 112-4041 112-4042 112-4043 112-4044
Rękawice nitrylowe odporne chemicznie, PowerCoat® 950-20 Neofit
Rękawice
This durable glove made from neoprene provides protection against a wide range of chemical products (detergents, some solvents, acids and bases).
  • Neoprene provides chemical and mechanical resistance
  • Diamond finish provides a better grip in damp environments
  • Cotton flock interior makes it easier to put on and off the glove
  • Long cuff helps to protect a large part of the forearm
Ideal for battery handling but can also be used to handle certain dilute acids.
Thickness: 0,72 mm
Colour: Black
EN 374 (AKL), EN 388 (3111), Cat. III
Certyfikaty: ENWięcej informacji o produkcie