Drukuj
Ochronne okulary, uvex 9161
Dostawca: UVEX
9161.005
111-8030EA 129 PLN
111-8030
Ochronne okulary, uvex 9161
Okulary
Cover glasses (OTG) with an unrestricted lateral field of vision can be worn over most prescription spectacles.
  • Indirect ventilation
  • Four position arm length adjustment
  • Ear piece pads guarantee an excellent, pressure-free fit
  • Optidur NHC coating for high scratch and chemical resistance
Weight: 56 g
EN 166-168, EN 170; 1F
Certyfikaty: EN 166, EN 170; 1F