Drukuj
Jednorazowe zatyczki do uszu, E-A-RSoft™
Dostawca: 3M
ES01001 ES01005 PD01010
111-1630EA 367 PLN
111-1630 111-1797 111-2422
Jednorazowe zatyczki do uszu, E-A-RSoft™
Zatyczki do uszu Zatyczki do uszu, jednorazowe
E-A-RSoft™ yellow neon earplugs are made from a slow expanding polyurethane foam material. A plug that gives evenly distributed pressure allows flexibility and a good seal with optimum comfort. Once inserted in the ear, the foam plugs expand to provide a snug and secure custom fit. These roll down earplugs are available with and without cord and be used with the One Touch™ dispenser.
  • Soft hypoallergenic foam material for lower pressure inside the ear
  • Smooth, dirt resistant surface for better hygiene, durability and comfort
  • Polyester cord helps prevent
Więcej informacji o produkcie